Learn More
Bio X Cell InVivoMab anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β, Clone: 1D11.16.8, Size: 50 MG
SDP
Supplier:  Bio X Cell BE0057R050MG
Encompass_Preferred
InVivoMab anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β, Clone: 1D11.16.8, Size: 50 MG, Isotype: mouse IgG1, Applications: in vivo TGFβ neutralization, in vitro TGFβ neutralization
Catalog No. 50-139-2846