Learn More
Bio X Cell InVivoPlus anti-mouse IFNγ, Clone: XMG
SDP
Supplier:  Bio X Cell BP0055R005MG
Encompass_Preferred
InVivoPlus anti-mouse IFNγ, Clone: XMG1.2, Size: 5 MG, Isotype: rat IgG1, Applications: in vivo IFNγ neutralization, in vitro IFNγ neutralization, ELISPOT, Flow cytometry
Catalog No. 50-139-3544