Learn More
Bio X Cell InVivoMab anti-mouse IL-6R, Clone: 15A7, Size: 25 MG
SDP
Supplier:  Bio X Cell BE0047R025MG
Encompass_Preferred
InVivoMab anti-mouse IL-6R, Clone: 15A7, Size: 25 MG, Isotype: rat IgG2b, Applications: in vivo blocking of IL-6/IL-6R signaling, in vitro blocking of IL-6R signaling
Catalog No. 50-139-2829