Learn More
Bio X Cell InVivoPlus anti-mouse OX40 (CD134), Clone: OX-86, Size: 5 MG
Supplier:  Bio X Cell BP0031R005MG
Encompass_Preferred
InVivoPlus anti-mouse OX40 (CD134), Clone: OX-86, Size: 5 MG, Isotype: rat IgG1, Applications: in vivo OX40 activation, in vitro OX40 activation
Catalog No. 50-139-3523