Learn More
Bio X Cell InVivoMab anti-mouse PD-1 (CD279), Clone: 29F.1A12, Size: 100 MG
SDP
Supplier:  Bio X Cell BE0273R100MG
Encompass_Preferred
InVivoMab anti-mouse PD-1 (CD279), Clone: 29F.1A12, Size: 100 MG, Isotype: rat IgG2a, kappa, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, Immunohistochemistry (frozen), Flow cytometry, Western blot
Catalog No. 50-139-3387