Learn More
Bio X Cell InVivoPlus anti-mouse PD-1 (CD279), Clone: J43, Size: 25 MG
SDP
Supplier:  Bio X Cell BP00332R025MG
Encompass_Preferred
InVivoPlus anti-mouse PD-1 (CD279), Clone: J43, Size: 25 MG, Isotype: Armenian hamster IgG, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization
Catalog No. 50-139-3530