Learn More
Bio X Cell InVivoPlus anti-mouse PD-L1 (B7-H1), Clone: 10F.9G2, Size: 100 MG
SDP
Supplier:  Bio X Cell BP0101R100MG
Encompass_Preferred
InVivoPlus anti-mouse PD-L1 (B7-H1), Clone: 10F.9G2, Size: 100 MG, Isotype: rat IgG2b, Applications: in vivo PD-L1 blockade, Immunofluorescence, Immunohistochemistry (frozen), Flow cytometry
Catalog No. 50-139-3592