Learn More
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Mouse anti-Mouse, PE, Clone: eBio25-D1.16 (25-D1.16), eBioscience
Mouse Monoclonal Antibody
Manufacturer: Invitrogen 12574381
Description
Description: The 25-D1.16 monoclonal antibody reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo. Applications Reported: This eBio25-D1.16 (25-D1.16) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio25-D1.16 (25-D1.16) antibody has been tested by flow cytometric analysis of SIINFEKL-pulsed C57Bl/6 splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Cells can be pulsed with the SIINFEKL peptide according to the following protocol: 1. With cells in flow staining buffer, add SIINFEKL peptide to a final concentration of 30 uM. 2. Incubate cells at 37°C for 2 hours. 3. Wash cells with flow staining buffer. 4. Proceed with cell surface staining as normal. For additional information see the references listed below. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.
2 µm post-manufacturing filtered.Specifications
OVA257-264 (SIINFEKL) peptide bound to H-2Kb | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
H-2Kb-SIINFEKL; OVA-Kb; OVA257-264 H-2Kb; OVA257-264 | |
IgG1, kappa | |
50 μg | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
Mouse |
Flow Cytometry | |
eBio25-D1.16 (25-D1.16) | |
PE | |
Liquid | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.