Learn More
anti-M13, HRP Conjugate, Monoclonal, GE Healthcare
Use for detection of M13 in a 96-well format. Anti-M13, HRP Conjugate, Monoclonal, GE Healthcare is perfect for ELISA assays.
Manufacturer:  GE Healthcare Life Sciences 27942101
Catalog No. 45-001-419
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Sufficient reagent for 200 96-well detection assays at 200µl/well (1:5000 dilution)
Host Species: Mouse
ELISA
Specifications
Specifications
M13
Ascites
1 Each
HRP
Murine