Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13 Phage coat protein, clone: E1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112900

Catalog No. 01-674-098

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.

Specifications

Specifications

M13 Phage coat protein
Monoclonal
1mg/mL
PBS with 0.08% sodium azide
Murine
IgG2a
1mg
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
Bacteria, Virus
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
E1
Unconjugated
Purified
M13 phage coat proteins.
Protein A/G
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit