Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A4A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524303

Catalog No. PIPA524303

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, rat and zebrafish based on sequence homology.

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. Alternative splicing of this gene results in several transcript variants; however, not all transcripts have been fully described.
Specifications

Specifications

MS4A4A
Polyclonal
Unconjugated
MS4A4A
Liquid
MS4A4A
KLH conjugated synthetic peptide between 37-64 amino acids from the N-terminal region of human M4A4A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q96JQ5
4SPAN1, CD20-L1, CD20L1, MS4A4, MS4A7, CD20 antigen-like 1, Fc epsilon receptor beta subunit homolog, four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51338
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit