Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP47, Polyclonal, Invitrogen™

Catalog No. PIPA127804
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127804 50 μg
1 options

Catalog No. PIPA127804

Supplier: Thermo Scientific PA127804

Chicken Polyclonal Antibody

Required for the transport of mannose 6-phosphate receptors (MPR) from endosomes to the trans-Golgi network. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein enco

PA1-27804 detects M6PRBP1 from human samples.PA1-27804 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Mannose-6-phosphate receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M6PRBP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene PLIN3
Gene Accession No. O60664
Gene Alias M6PRBP1, MGC11117, MGC2012, PP17, TIP47
Gene Symbols PLIN3
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Mannose-6-phosphate receptor.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.