Learn More
anti-Macro H2A.2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232328
Catalog No. NBP232328
Add to cart
Description
Description
Macro H2A.2 Polyclonal antibody specifically detects Macro H2A.2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Macro H2A.2
0.5 mg/mL
Western Blot 1:1000
Q9P0M6
core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2
Rabbit
IgG
Protein A purified
RUO
Primary
55506
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
H2AFY2
This H2AFY2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 148-169 amino acids from the Central region of human H2AFY2.
40.058 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human