Learn More
MAD1p Rabbit anti-Yeast, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA15831
Catalog No. 01-674-979
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-5831 detects Mad1p in yeast samples. PA1-5831 has been successfully used in Western blot and immunofluorescence procedures. The PA1-5831 immunogen is purified recombit GST-Mad1p.
Specifications
Specifications
Mad1p
Polyclonal
whole serum with 0.1% sodium azide
Liquid
MAD1
Purified recombit GST-Mad1p.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
852794
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MAD1
YGL086W
Rabbit
IgG
RUO
Antibody
Polyclonal
Yeast