Learn More
MAD2A Mouse anti-Human, Rat, Mouse, Clone: 17D10, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051501
Catalog No. 05-150-1MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-MAD2A Antibody, clone 17D10 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation to detect MAD2A also known as MAD2 mitotic arrest deficient-like protein 1 (yeast).
Specifications
Specifications
MAD2A
17D10
Purified mouse monoclonal in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
Q13257
HSMAD2, HsMAD2, MAD2
Recombinant protein corresponding to full length human MAD2A.
Protein G purfied
2-8°C for up to 12 months.
Monoclonal