Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2B/Rev7 Mouse, Unlabeled, Clone: 14, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612266

Catalog No. BDB612266


Add to cart

Description

Description

Progression of the mammalian cell cycle is regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins. These events are of utmost importance at the checkpoints, or transition points, of the cell cycle. MAD2 (Mitotic Arrest-Deficient) is the human homolog of a yeast and Xenopus protein that is essential for spindle assembly during mitosis. Binding of affinity purified polyclonal antibodies to the MAD2 protein has been reported to prevent mitosis of HeLa cells. Furthermore, MAD2 is localized at the kinetochore of condensed chromosomes during mitosis and cells defective in the mitotic checkpoint have reduced levels of MAD2. MAD2 inhibits anaphase-promoting complex (APC) ubiquitin ligase activity,which is critical for controlling transitions in mitosis. This inhibition occurs through binding and suppression of CDC20 activation of APC. MAD2B, also known as Rev7, is a UV revertible gene that has 25% identity with MAD2 and also acts as an inhibitor of APC. Interestingly, MAD2B/Rev7 can bind MAD2 and interacts with the DNA polymerase ζ subunit Rev3. Thus, MAD2B may interact with mitotic checkpoint proteins and DNA repair proteins to regulate mitotic progression.

Immunofluorescence, Western Blotting

Specifications

Specifications

MAD2B/Rev7
14
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Mitotic Arrest Deficient-2B; Rev7
Affinity Purified
Human MAD2B aa. 81-180
50μg
Cell Biology
Primary
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit