Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1BP, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127838

Catalog No. PIPA127838

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May function to silence the spindle checkpoint and allow mitosis to proceed through anaphase by binding MAD2L1 after it has become dissociated from the MAD2L1-CDC20 complex. The protein encoded by this gene was identified as a binding protein of the MAD2

PA1-27838 detects MAD2L1BP from human samples.PA1-27838 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human MAD2L1-binding protein.
Specifications

Specifications

MAD2L1BP
Polyclonal
PBS with no preservative
Q15013
CMT2, KIAA0110, MGC11282, RP1-261G23.6
Chicken
IgY
50μg
Store at -20°C
Primary
9587
Western Blot
Unconjugated
MAD2L1BP
Purified
MAD2L1BP
Synthetic peptide corresponding to human MAD2L1-binding protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit