Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131766

Catalog No. PA131766


Add to cart

Description

Description

PA1-31766 detects MAD2L2 from human samples. PA1-31766 has been successfully used in Western blot applications. The PA1-31766 immunogen is: Synthetic peptide: TLTRQDLNFGQV, corresponding to amino acids 3-14 of Human Mad2L2.

The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins.
Specifications

Specifications

MAD2L2
Polyclonal
Unconjugated
MAD2L2
Liquid
MAD2L2
synthetic peptide TLTRQDLNFGQV, corresponding to amino acids 3-14 of Human Mad2L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.81 mg/mL
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Q9UI95
RP3-330O12.4, MAD2B, REV7
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10459
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit