Learn More
MAD2p Rabbit anti-Yeast, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA15834
Catalog No. 01-674-980
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-5834 detects Mad2p in yeast samples. PA1-5834 has been successfully used in Western blot and immunofluorescence procedures. The PA1-5834 immunogen is purified bacterial recombit His6-fusion Mad2p.
mad2 is required for the execution of the mitotic checkpoint which monitors the process of kinetochore-spindle attachment and delays the onset of anaphase when this process is not complete. It inhibits the activity of the anaphase promoting complex by sequestering cdc20 until all chromosomes are aligned at the metaphase plate.
Specifications
Specifications
Mad2p
Polyclonal
whole serum with 0.1% sodium azide
Liquid
MAD2
Purified bacterial recombit His6-fusion Mad2p.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
853422
Immunofluorescence, Western Blot
Unconjugated
MAD2
YGL086W
Rabbit
IgG
RUO
Antibody
Polyclonal
Yeast