Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAFA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526449

Catalog No. PIPA526449

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene.
Specifications

Specifications

MAFA
Polyclonal
PBS with 0.09% sodium azide
Q8NHW3
RIPE3b1, hMafA, pancreatic beta-cell-specific transcriptional activator, transcription factor MafA, transcription factor RIPE3b1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389692
Western Blot
Unconjugated
MAFA
Liquid
MAFA
KLH conjugated synthetic peptide between 282-311 amino acids from the C-terminal region of human MAFA
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit