Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP217203

Catalog No. NBP217203


Add to Cart

Description

Description

MAGEA3 Polyclonal specifically detects MAGEA3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MAGEA3
Polyclonal
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613
Rabbit
35 kDa
0.1 mL
Cancer
4102
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
1mg/mL
Western Blot 1:500-1:3000
P43357
MAGEA3
Recombinant protein encompassing a sequence within the center region of human MAGEA3. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Human
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only