Learn More
MAGI3 Mouse anti-Rat, Clone: SLP-32, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA125158
Description
MA1-25158 detects MAGI3 from rat samples. MA1-25158 has been successfully used in immunocytochemistry, and Western blot procedures. The MA1-25158 immunogen is a synthetic peptide corresponding to C-terminal residues 421-785 of rat MAGI3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
associates with tyrosine-phosphorylated proteins to link the receptor with its substrate in the plasma membrane.Specifications
MAGI3 | |
Monoclonal | |
1.7 mg/mL | |
Magi3 | |
Slipr | |
Mouse | |
Protein A | |
RUO | |
245903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
SLP-32 | |
Unconjugated | |
Q9JK71 | |
MAGI3 | |
Synthetic peptide corresponding to C-terminal residues 421-785 of rat MAGI3. | |
50 μL | |
Primary | |
Rat | |
Antibody | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.