Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGI3 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125159

Catalog No. PIPA125159


Add to cart

Description

Description

PA1-25159 detects MAGI3 from rat samples. PA1-25159 has been successfully used in Western blot procedures. The PA1-25159 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 835-850 of rat MAGI3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

associates with tyrosine-phosphorylated proteins to link the receptor with its substrate in the plasma membrane.
Specifications

Specifications

MAGI3
Polyclonal
Unconjugated
Magi3
Liquid
MAGI3
Synthetic peptide, conjugated to KLH, corresponding to residues 835-850 of rat MAGI3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Immunocytochemistry, Immunofluorescence, Western Blot
0.8 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
Q9JK71
Slipr
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
245903
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit