Learn More
male germ cell-associated kinase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MAK protein.
Supplier: Abnova Corporation H00004117D01P
Description
The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized. [provided by RefSeq
Sequence: MNRYTTMRQLGDGTYGSVLMGKSNESGELVAIKRMKRKFYSWDECMNLREVKSLKKLNHANVIKLKEVIRENDHLYFIFEYMKENLYQLMKDRNKLFPESVIRNIMYQILQGLAFIHKHGFFHRDMKPENLLCMGPELVKIADFGLARELRSQPPYTDYVSTRWYRAPEVLLRSSVYSSPIDVWAVGSIMAELYMLRPLFPGTSEVDEIFKICQVLGTPKKSDWPEGYQLASSMNFRFPQCVPINLKTLIPNASNEAIQLMTEMLNWDPKKRPTASQALKHPYFQVGQVLGPSSNHLESKQSLNKQLQPLESKPSLVEVEPKPLPDIIDQVVGQPQPKTSQQPLQPIQPPQNLSVQQPPKQQSQEKPPQTLFPSIVKNMPTKPNGTLSHKSGRRRWGQTIFKSGDSWEELEDYDFGASHSKKPSMGVFKEKRKKDSPFRQQVKMAVISLSAHQFPTLSpecifications
male germ cell-associated kinase | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human MAK protein. | |
MAK | |
dJ417M14.2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC039825.1 | |
MAK | |
MAK (AAH39825.1, 1 a.a. ∼ 457 a.a) full-length human protein. | |
100 μg | |
Kinases | |
4117 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.