Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARVELD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526228

Catalog No. PIPA526228

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MALD2 plays a role in the formation of the epithelial barriers. The separation of the endolymphatic and perilymphatic spaces of the organ of Corti from one another by epithelial barriers is required for normal hearing.
Specifications

Specifications

MARVELD2
Polyclonal
PBS with 0.09% sodium azide
Q8N4S9
DFNB49, MARVD2, MRVLDC2, Tric, MARVEL (membrane-associating) domain containing 2, MARVEL domain-containing protein 2, tricellulin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
153562
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MARVELD2
Liquid
MARVELD2
KLH conjugated synthetic peptide between 386-415 amino acids from the C-terminal region of human MALD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit