Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Receptor Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14111

Catalog No. PA14111


Add to cart

Description

Description

PA1-4111 detects mouse mammary tumor virus receptor in mouse samples. PA1-4111 has been successfully used in Western blot procedures. The PA1-4111 immunogen is a synthetic peptide corresponding to residues D(152) D E E P P D A S L P P C(163) of the 20 kDa MMTV-R.

MMTV is a retrovirus, which is a causative agent of mammary carcinomas in vivo. The MMTV receptor (MMTV-R) is found in humans and other mammals.
Specifications

Specifications

Transferrin Receptor
Polyclonal
Unconjugated
Tfrc
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
22042
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q62351
TFRC
Synthetic peptide corresponding to residues D(152) D E E P P D A S L P P C(163) of the 20 kDa MMTV-R.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit