Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAMSTR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524109

Catalog No. PIPA524109

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcriptional coactivator. Stimulates the transcriptional activity of MEF2C. Stimulates MYOD1 activity in part via MEF2, resulting in an enhancement of skeletal muscle differentiation.
Specifications

Specifications

MAMSTR
Polyclonal
PBS with 0.09% sodium azide
Q6ZN01
MASTR, MEF2-activating SAP transcriptional regulator, MEF2-activating SAP transcriptional regulatory protein, MEF2-activating motif and SAP domain-containing transcriptional regulator, likely ortholog of MEF2-activating SAP transcriptional regulator
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284358
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MAMSTR
Liquid
MAMSTR
KLH conjugated synthetic peptide between 9-37 amino acids from the N-terminal region of human MAMSTR
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit