Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN2A1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524447

Catalog No. PIPA524447

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein which is a member of family 38 of the glycosyl hydrolases. The protein is located in the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus.
Specifications

Specifications

MAN2A1
Polyclonal
PBS with 0.09% sodium azide
Q16706
GOLIM7, MANA2, MANII, AMan II, Golgi alpha-mannosidase II, alpha-mannosidase 2, golgi integral membrane protein 7, man II, mannosidase, alpha type II, mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4124
Flow Cytometry, Western Blot
Unconjugated
MAN2A1
Liquid
MAN2A1
KLH conjugated synthetic peptide between 456-482 amino acids from the central region of human MAN2A1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit