Learn More
MAP Kinase 2/Erk2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06333
Catalog No. 06-333-MI
Add to cart
Description
Description
Anti-MAP Kinase 2/Erk2 Antibody detects level of MAP Kinase 2/Erk2 and has been published and validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
MAP Kinase 2/Erk2
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_002745
Immunoprecipitation, Western Blot
0.02M phosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide
MAPK1; p38; ERK; P42MAPK; ERK2; p40; PRKM1; p41mapk; PRKM2; MAPK2; p42-MAPK; p41; ERK-2; ERT1
Recombinant mouse ERK2
Protein G chromatography
−20°C for 24 months
Polyclonal
Human, Murine, Rat
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021