Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14269

Catalog No. PA14269


Add to cart

Description

Description

PA1-4269 detects the human MAP126 protein in human samples. PA1-4269 has been successfully used in Western blot procedures. PA1-4269 immunizing peptide corresponds to amino acid residues 532-543 of human MAP126 protein.

A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.
Specifications

Specifications

SPAG5
Polyclonal
Unconjugated
SPAG5
Liquid
SPAG5
Synthetic peptide: V(532) N Q E S R R A E T L V(543)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96R06
Sperm-associated antigen 5, Astrin, Mitotic spindle-associated protein p126
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10615
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit