Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1B Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612678

 View more versions of this product

Catalog No. BDB612678


Add to cart

Description

Description

Microtubule-associated proteins (MAPs) play a crucial role in the development and structure of nerve cells. These proteins are important for the assembly and stability of microtubles during neurite outgrowth and for the morphology of neuronal processes, such as dendrites. MAP1B encodes for a single precursor protein that is cleaved to produce a single heavy chain of 280kDa and a light chain of 32kDa which become noncovalently associated. Expression levels of MAP1B are reportedly highest in the brain and are modulated during development as reported with gene silencing experiments that implicate MAP1B's role during neuronal differentiation. In addition, MAP1B can be selectively phosphorylated depending on the cell type, subcellular localization and developmental phase. In neurons, MAP1B is expressed as a glycoprotein at the plasma membrane, where it can interact with proteins participating in axonal guiding. MAP1B has been reported to be observable to migrate in a range between 320-340kDa.

Western Blotting

Specifications

Specifications

MAP1B
6
Unconjugated
Affinity Purified
Mouse MAP1B aa. 1745-1858
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.