Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAP1B (p Thr1265), Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142827AF647

Catalog No. NBP142827Y


Add to cart

Description

Description

MAP1B p Thr1265 Polyclonal antibody specifically detects MAP1B p Thr1265 in Human, Mouse, Rat, Chicken, Fish, Reptile, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

MAP1B (p Thr1265)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Phosphorylated Thr1265 on MAP1B (Thr1261 in rat).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Affinity Purified
MAP1B
Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot# P15205]
320 kDa
0.1 ml
Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers
Primary
4131
Chicken, Fish, Human, Mouse, Rat, Reptile, Xenopus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only