Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1B, p Thr1265 Antibody, FITC - SuperBUGS, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142827F
Description
MAP1B Polyclonal specifically detects MAP1B in Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
MAP1B (p Thr1265) | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B | |
Rabbit | |
320 kDa | |
0.1 mL | |
Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers | |
4131 | |
Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus species | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
FITC | |
PBS with 0.05% Sodium Azide | |
MAP1B | |
Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot# P15205] | |
Affinity Purified | |
RUO | |
Primary | |
Phosphorylated Thr1265 on MAP1B (Thr1261 in rat). | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction