Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METAP1D Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524339

Catalog No. PIPA524339

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
Specifications

Specifications

METAP1D
Polyclonal
PBS with 0.09% sodium azide
Q9CPW9, Q6UB28
MAP1D, Metap1l, CDS of metAP-3 within PCR fragment, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
254042, 66559
Western Blot
Unconjugated
METAP1D
Liquid
METAP1D
KLH conjugated synthetic peptide between 306-334 amino acids from the N-terminal region of human MAP1D
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit