Learn More
LC3A Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OSA00003W
Catalog No. PIOSA00003W
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 150 µL of distilled water. Centrifuge to remove any insoluble material.
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. ; This protein is probably involved in formation of autophagosomal vacuoles. Most abundant in heart, brain, liver, skeletal muscle and testis but absent in thymus and peripheral blood leukocytes.
Specifications
Specifications
LC3A
Polyclonal
whole serum with no preservative
Q9H492, Q91VR7, Q6XVN8
MAP1A/MAP1B LC3 A, MAP1A/1B light chain 3 A, MAP1 light chain 3-like protein 1, Microtubule-associated protein 1 light chain 3 alpha, Autophagy-relate
Sheep
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
MAP1LC3A
Lyophilized
MAP1LC3A
A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3A
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
362245, 66734, 84557