Learn More
LC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSM00024W
Catalog No. PIOSM00024W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 150 μL of distilled water.
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LC3A
Polyclonal
MAP1LC3A
MAP1ALC3, MAP1BLC3, MAP1A/MAP1B LC3 A, microtubule-associated protein 1 light chain 3 alpha
Rabbit
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9H492, Q91VR7, Q6XVN8
MAP1LC3A
A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3A
RUO
362245, 66734, 84557
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations