Learn More
MAP1S Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525672
Catalog No. PIPA525672
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule-organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.
Specifications
Specifications
MAP1S
Polyclonal
Unconjugated
MAP1S
Liquid
MAP1S
KLH conjugated synthetic peptide between 493-520 amino acids from the central region of human MAP1S
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q66K74
BPY2IP1, C19orf5, MAP8, VCY2IP-1, VCY2IP1, BPY2 interacting protein 1, BPY2-interacting protein 1, MAP-1S, VCY2 interacting protein 1, VCY2-interacting protein 1, microtubule-associated protein 8, variable charge Y chromosome 2-interacting protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55201