Learn More
MAP2 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSM00030W
Catalog No. PIOSM00030W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and maligt brain tissue as well as in degenerative diseases such as Alzheimer ft.s Disease (AD).
Specifications
Specifications
MAP2
Polyclonal
whole serum with no preservative
P20357, P15146
Microtubule-associated protein 2, MAP 2, MAP-2, MAP2R
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Map2
Lyophilized
MAP2
A synthetic peptide from mouse MAP2 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17756, 25595