Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524589

Catalog No. PIPA524589

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MAP2 encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dentrites, implicating a role in determining and stabilizing dentritic shape during neuron development.

Specifications

Specifications

MAP2
Polyclonal
PBS with 0.09% sodium azide
P11137, P20357
MAP2A, MAP2B, MAP2C, MAP-2
Rabbit
IgG
400μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17756, 4133
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MAP2
Liquid
MAP2
MAP2 recombinant protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit