Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2B Mouse, Unlabeled, Clone: 18/MAP2B, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610460

Catalog No. BDB610460


Add to cart

Description

Description

Microtubule-associated proteins (MAPs) play a crucial role in the development and morphology of neurons. MAP2, specifically localized in dendrites, has four known isoforms that are produced by alternative splicing of the transcript and are expressed at various stages of neuronal development. MAP2B is a 280-kDa protein that is expressed throughout brain development. It contains functional domains that interact with the regulatory subunit of the cAMP-dependent kinase II and microtubules. Experimental monitoring of its presence, along with GFAP and nestin, may be utilized to quantify neuronal development.

Specifications

Specifications

MAP2B
18/MAP2B
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit