Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mitogen-activated protein kinase kinase 7 (A01), Mouse anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant MAP2K7.

Manufacturer:  Abnova Corporation H00005609A01

Catalog No. 89-004-263


Add to cart

Description

Description

The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase specifically activates MAPK8/JNK1 and MAPK9/JNK2, and this kinase itself is phosphorylated and activated by MAP kinase kinase kinases including MAP3K1/MEKK1, MAP3K2/MEKK2,MAP3K3/MEKK5, and MAP4K2/GCK. This kinase is involved in the signal transduction mediating the cell responses to proinflammatory cytokines, and environmental stresses. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found, but only one transcript variant has been supported and defined. [provided by RefSeq

Sequence: MAASSLEQKLSRLEAKLKQENREARRRIDLNLDISPQRPRPTLQLPLANDGGSRSPSSESSPQHPTPPARPRHMLGLPSTLFTPRSMESIEIDQKLQEI
Specifications

Specifications

mitogen-activated protein kinase kinase 7
Unconjugated
50% glycerol
NM_145185
Jnkk2|MAPKK7|MKK7|PRKMK7
Mouse
50 uL
Kinases
Y
Polyclonal
Human, Murine, Rat
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant MAP2K7.
MAP2K7
Serum
MAP2K7
MAP2K7 (NP_660186, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5609
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit