Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137640

Catalog No. 01-673-423


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, canine, amphibian, porcine, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is breast carcinoma.

ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
Specifications

Specifications

ERK2
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P28482, P63085
ERK, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, ERK-2, MAP kinase 1, MAP kinase 2, MAP kinase isoform p42, MAPK 2, extracellular signal-regulated kinase 2, mitogen-activated protein kinase 2, p42-MAPK, protein tyrosine kinase ERK2
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26413, 5594
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
MAPK1
Liquid
MAPK1
Synthetic peptide derived from the C-terminus of mouse ERK-2 protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit