Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mitogen-activated protein kinase 15 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAPK15 protein.
Manufacturer: Abnova Corporation H00225689B02
Description
Sequence: MCTVVDPRIVRRYLLRRQLGQGAYGIVWKAVDRRTGEVVAIKKIFDAFRDKTDAQRTFREITLLQEFGDHPNIISLLDVIRAENDRDIYLVFEFMGCPPSPPPPTAVRTLSADTDLNAVIRKGGLLQDVHVRSIFYQLLRATRFLHSGHVVHRDQKPSNVLLDANCTVKLCDFGLARSLGDLPEGPEDQAVTEYVATRWYRAPEVLLSSHRYTLGVDMWSLGCILGEMLRGRPLFPGTSTLHQLELILETIPPPSEEDLLALGSGCRASVLHQLGSRSpecifications
mitogen-activated protein kinase 15 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MAPK15 protein. | |
MAPK15 | |
ERK7/ERK8 | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
No additive | |
BC028034.1 | |
MAPK15 | |
MAPK15 (AAH28034.1, 1 a.a. ∼ 277 a.a) full-length human protein. | |
RUO | |
Yes | |
225689 | |
Serum |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title