Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1IP1L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526735

Catalog No. PIPA526735

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and non-human primate based on sequence homology.

Specifications

Specifications

MAPK1IP1L
Polyclonal
PBS with 0.09% sodium azide
Q8NDC0
C14orf32, MISS, c14_5346, MAPK-interacting and spindle-stabilizing protein, MAPK-interacting and spindle-stabilizing protein-like, mitogen activated protein kinase 1 interacting protein 1-like, mitogen-activated protein kinase 1-interacting protein 1-like
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
93487
Immunohistochemistry, Western Blot
Unconjugated
MAPK1IP1L
Liquid
MAPK1IP1L
KLH conjugated synthetic peptide between 108-137 amino acids from the central region of human MAPK1IP1L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit