Learn More
MAPK3 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of MAPK3.
Manufacturer: Abnova Corporation PAB16332
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described. [provided by RefSeq
Specifications
MAPK3 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7 ug/mL) The optimal working dilution should be determined by the end user. | |
MAPK3 | |
ERK1|HS44KDAP|HUMKER1A|MGC20180|P44ERK1|P44MAPK|PRKM3 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
Primary | |
5595 | |
Bovine, Canine, Equine, Hamster, Human, Monkey, Murine, Porcine, Rat |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of MAPK3. | |
In PBS (0.09% sodium azide) | |
Liquid | |
MAPK3 | |
A synthetic peptide (conjugated with KLH) corresponding to human MAPK3. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Internal domain of human. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only