Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK5, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127890

Catalog No. PIPA127890

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MEK5 and ERK5 interact specifically with one another and not with MEK1/ERK1 or MEK2/ERK2 pathways. The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals. and are inv

PA1-27890 detects MAPK7 from human samples.PA1-27890 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Mitogen-activated protein kinase 7.
Specifications

Specifications

ERK5/MAPK7
Polyclonal
PBS with no preservative
Q13164
BMK1, ERK4, ERK5, PRKM7
Chicken
IgY
50μg
Store at -20°C
Primary
5598
Western Blot
Unconjugated
MAPK7
Purified
MAPK7
Synthetic peptide corresponding to human Mitogen-activated protein kinase 7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit