Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141234

Catalog No. 01-675-588


Add to cart

Description

Description

Suggested positive control: A-431 whole cell lysate1.

c-Jun amino terminal kinases (JNKs) are MAP kinase family members that differ from other MAP kinases in that they are phosphorylated at the Thr-Pro-Tyr phosphorylation motif instead of the characteristic MAP kinase Thr-Glu-Tyr motif. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
Specifications

Specifications

JNK2
Polyclonal
Unconjugated
MAPK9
Liquid
MAPK9
Synthetic peptide corresponding to residues 373-389 K(373) D Q P S D A A V S S N A T P S Q(389) of human JNK2.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
P45984
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, p54a, p54aSAPK
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5601
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit