Learn More
MAPKAP-K2 Mouse anti-Human, Mouse, Rat, Clone: 7H4.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 04539
Catalog No. 04-539-MI
Add to Cart
Description
Description
Anti-MAPKAP-K2 Antibody, clone 7H4.2 detects level of MAPKAP-K2 and has been published and validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
MAPKAP-K2
Monoclonal
Unconjugated
NM_004759.3; NM_032960.2
Mouse
Protein A purfied
Primary
−20°C for 24 months.
IgG
Immunoprecipitation, Western Blot
7H4.2
200μg (1mg/mL) of mouse monoclonal IgG in PBS containing 0.1% sodium azide.
MAPKAPK2, MK2, MAPKAPK-2
KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2.
200 μg
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions