Learn More
MAPKAPK2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA124383
Catalog No. PIPA124383
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The MAPKAP kinases (MAP kinase activated protein kinases) are a group of MAP kinase substrates that also have kinase activity. This family of catalytic proteins includes Rsk-1 (also known as MAPKAP Kinase-1), as well as MAPKAP Kinase-2 and MAPKAP Kinase-3 (also known as 3pK), each of which functions as an intermediate kinase in a cell signaling pathway.
PA1-24383 detects MAPKAP Kinase2 in Human samples.PA1-24383 has been successfully used in Western Blot procedures.PA1-24383 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human MAPKAPK2.
Specifications
Specifications
MAPKAPK2
Polyclonal
Unconjugated
MAPKAPK2
Purified
MAPKAPK2
Synthetic peptide region of human MAPKAPK2.
Protein A/G
RUO
Antibody
Polyclonal
Human
Western Blot
1mg/mL
PBS with 2% sucrose and no preservative
P49137
MK2
Rabbit
IgG
100μg
Avoid Freeze/Thaw Cycles
Primary
9261