Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAPK2 Rabbit anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126428

Catalog No. PA126428


Add to cart

Description

Description

PA1-26428 detects MAPKAP Kinase 2 from bovine, canine, human, mouse, and rat samples. PA1-26428 has been successfully used in Western blot procedures. The PA1-26428 immunogen is considered to be commercially sensitive.

This gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

MAPKAPK2
Polyclonal
Unconjugated
MAPKAPK2
Liquid
MAPKAPK2
This information is considered to be commercially sensitive.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Rat
Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 1% BSA and 0.02% sodium azide
P49137, P49138
MK2
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17164, 289014, 490263, 788091, 9261
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit